Assay Details
| Gene Symbol: MIR1840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049316.1 | 1 | 32 | 56 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin Pas1/PHO80 domain containing 1 | CNPPD1 | Anti-sense | Chr. 37 - 25729196 - 25734724 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR1840
Genomic Map