Assay Details
| Gene Symbol: MIR92B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049345.1 | 1 | 35 | 49 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calpain 8 | CAPN8 | Anti-sense | Chr. 7 - 40408408 - 40451843 on NCBI Build CanFam3.1 | ||
| Downstream | Fc receptor-like protein 2 | LOC490399 | 45.5kb | Sense | Chr. 7 - 40479954 - 40491160 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR92B
Genomic Map