Assay Details
Gene Symbol: MIR496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049495.1 | 1 | 47 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | VRK serine/threonine kinase 1 | VRK1 | -1.6mb | Sense | Chr. 8 - 65444404 - 65551348 on NCBI Build CanFam3.1 | |
Downstream | BAF chromatin remodeling complex subunit BCL11B | BCL11B | 493.2kb | Anti-sense | Chr. 8 - 67622592 - 67713922 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
