Assay Details
| Gene Symbol: MIR505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049497.1 | 1 | 39 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MCF.2 cell line derived transforming sequence | MCF2 | -143.6kb | Sense | Chr. X - 109543609 - 109627265 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR505
Genomic Map