Assay Details
| Gene Symbol: MIR8893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128859.1 | 1 | 56 | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BCL2 associated transcription factor 1 | BCLAF1 | -167.1kb | Anti-sense | Chr. 1 - 28852654 - 28882095 on NCBI Build CanFam3.1 | |
| Downstream | mitogen-activated protein kinase kinase kinase 5 | MAP3K5 | 58.5kb | Anti-sense | Chr. 1 - 29107813 - 29306437 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8893
Genomic Map