Assay Details
| Gene Symbol: MIR652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049393.1 | 1 | 25 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102152628 | LOC102152628 | -148.9kb | Sense | Chr. X - 83022557 - 83027457 on NCBI Build CanFam3.1 | |
| Downstream | transmembrane protein 164 | TMEM164 | 43.1kb | Sense | Chr. X - 83219530 - 83384900 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR652
Genomic Map