Assay Details
| Gene Symbol: MIR8796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128736.1 | 1 | 44 | 108 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
nuclear factor, erythroid 2 like 2 |
NFE2L2 | -28.3kb | Sense | Chr. 36 - 20989205 - 21087044 on NCBI Build CanFam3.1 | |
| Downstream | alkylglycerone phosphate synthase | AGPS | 16.1kb | Anti-sense | Chr. 36 - 21131616 - 21264615 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8796
Genomic Map