Assay Details
| Gene Symbol: MIR8798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128738.1 | 1 | 76 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uridine-cytidine kinase 2 | UCK2 | -23.1kb | Sense | Chr. 38 - 17475789 - 17555926 on NCBI Build CanFam3.1 | |
| Downstream | transmembrane and coiled-coil domains 1 | TMCO1 | 350.0 | Anti-sense | Chr. 38 - 17579540 - 17630701 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8798
Genomic Map