Assay Details
| Gene Symbol: MIR8828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128771.1 | 1 | 65 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Mdm1 nuclear protein | MDM1 | -71.2kb | Sense | Chr. 10 - 10523656 - 10552824 on NCBI Build CanFam3.1 | |
| Downstream |
RAP1B, member of RAS oncogene family |
RAP1B | 181.2kb | Anti-sense | Chr. 10 - 10805359 - 10816839 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8828
Genomic Map