Assay Details
| Gene Symbol: MIR8858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128809.1 | 1 | 94 | 120 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | apoptosis inhibitor 5 | API5 | -19.7kb | Sense | Chr. 18 - 26326387 - 26355580 on NCBI Build CanFam3.1 | |
| Downstream | leucine rich repeat containing 4C | LRRC4C | 1.3mb | Anti-sense | Chr. 18 - 27687197 - 28851487 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8858
Genomic Map