Assay Details
| Gene Symbol: MIR8882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128841.1 | 1 | 94 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
delta 4-desaturase, sphingolipid 2 |
DEGS2 | -13.2kb | Sense | Chr. 8 - 68461461 - 68483685 on NCBI Build CanFam3.1 | |
| Downstream | YY1 transcription factor | YY1 | 42.4kb | Anti-sense | Chr. 8 - 68539511 - 68572796 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8882
Genomic Map