Assay Details
| Gene Symbol: MIR8884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128843.1 | 1 | 66 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurexin-3 | LOC490812 | NRXN3 | -54.2kb | Anti-sense | Chr. 8 - 50848045 - 51609368 on NCBI Build CanFam3.1 |
| Downstream | neurexin-3-beta | LOC106557488 | 543.1kb | Anti-sense | Chr. 8 - 52206774 - 52386317 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8884
Genomic Map