Assay Details
| Gene Symbol: MIR449B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128867.1 | 1 | 81 | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myosin IIIA | MYO3A | -179.3kb | Sense | Chr. 2 - 7455125 - 7695483 on NCBI Build CanFam3.1 | |
| Downstream | G protein-coupled receptor 158 | GPR158 | 94.9kb | Sense | Chr. 2 - 7969781 - 8379657 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR449B
Genomic Map