Assay Details
| Gene Symbol: MIR506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128884.1 | 1 | 75 | 110 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA polymerase II elongation factor ELL2-like | LOC102152097 | -405.2kb | Anti-sense | Chr. X - 115036815 - 115151635 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR506
Genomic Map