Assay Details
| Gene Symbol: cOR9S7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003184.1 | 1 | 179 | 91 | ||
| GenBank mRNA | X89664.1 | - | 179 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 169 | CCDC169 | Sense | Chr. 25 - 4290543 - 4334981 on NCBI Build CanFam3.1 | ||
| Downstream | spermatogenesis and oogenesis specific basic helix-loop-helix 2 | SOHLH2 | SOSF2 | 36.1kb | Sense | Chr. 25 - 4345785 - 4419352 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

cOR9S7P
Genomic Map