Assay Details
| Gene Symbol: NME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024637.1 | NP_001019808.1 | 1 | 224 | 74 | |
| XM_005624340.3 | XP_005624397.1 | 1 | 386 | 74 | ||
| XM_005624341.3 | XP_005624398.1 | 1 | 353 | 74 | ||
| GenBank mRNA | AB207044.1 | - | 1 | 224 | 74 | |
| DN372571.1 | - | 1 | 280 | 74 | ||
More Information
Set Membership: |
|

NME1
Genomic Map