Assay Details
| Gene Symbol: RPL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252145.1 | NP_001239074.1 | 4 - 5 | 346 | 101 | |
| XM_005627055.3 | XP_005627112.1 | 4 - 5 | 344 | 101 | ||
| GenBank mRNA | GR887142.1 | - | 4 - 5 | 346 | 101 | |
More Information
Set Membership: |
|

RPL10A
Genomic Map