Assay Details
| Gene Symbol: TIMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284439.1 | NP_001271368.1 | 1 - 2 | 128 | 58 | |
| GenBank mRNA | JF508171.1 | - | 1 - 2 | 128 | 58 | |
| JX144398.1 | - | 1 - 2 | 54 | 58 | ||
More Information
Set Membership: |
|

TIMP3
Genomic Map