Assay Details
| Gene Symbol: DEFB119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284444.1 | NP_001271373.1 | 1 - 2 | 53 | 64 | |
| GenBank mRNA | DQ011989.1 | - | 1 - 2 | 53 | 64 | |
More Information
Set Membership: |
|

DEFB119
Genomic Map