Assay Details
| Gene Symbol: MAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284493.1 | NP_001271422.1 | 211 | 96 | ||
| GenBank mRNA | HQ693063.1 | - | 211 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001284493.1 transcripts.
Set Membership: |
|

MAS1
Genomic Map