Assay Details
| Gene Symbol: PROCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314130.1 | NP_001301059.1 | 2 - 3 | 333 | 107 | |
| XM_005634966.3 | XP_005635023.1 | 3 - 4 | 509 | 107 | ||
| XM_005634967.2 | XP_005635024.1 | 3 - 4 | 462 | 107 | ||
| GenBank mRNA | KF498853.1 | - | 2 - 3 | 333 | 107 | |
More Information
Set Membership: |
|

PROCR
Genomic Map