Assay Details
| Gene Symbol: KRT26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346066.1 | NP_001332995.1 | 1005 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KRT26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346066.1 | NP_001332995.1 | 1005 | 76 | ||