Assay Details
| Gene Symbol: LOC106557476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314090.1 | NP_001301019.1 | 1 - 2 | 103 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001314090.1 transcripts.
Set Membership: |
|

LOC106557476
Genomic Map