Assay Details
| Gene Symbol: Tead3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649256.1 | XP_007647446.1 | 9 - 10 | 967 | 68 | |
| XM_007649257.1 | XP_007647447.1 | 9 - 10 | 938 | 68 | ||
More Information
Set Membership: |
|

Tead3
Genomic Map