Assay Details
| Gene Symbol: Kat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641219.1 | XP_007639409.1 | 15 - 16 | 1986 | 91 | |
| GenBank mRNA | JI882115.1 | - | 3 - 4 | 299 | 91 | |
| JP043709.1 | - | 3 - 4 | 299 | 91 | ||
More Information
Set Membership: |
|

Kat2b
Genomic Map