Assay Details
| Gene Symbol: Mpv17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497050.2 | XP_003497098.2 | 6 - 7 | 524 | 74 | |
| GenBank mRNA | JI880701.1 | - | 6 - 7 | 446 | 74 | |
| JP047455.1 | - | 6 - 7 | 443 | 74 | ||
More Information
Set Membership: |
|

Mpv17
Genomic Map