Assay Details
| Gene Symbol: Sumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500595.2 | XP_003500643.1 | 4 - 5 | 463 | 60 | |
| GenBank mRNA | JP059569.1 | - | 1 - 2 | 118 | 60 | |
More Information
Set Membership: |
|

Sumo3
Genomic Map