Assay Details
| Gene Symbol: Wbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506817.2 | XP_003506865.1 | 7 - 8 | 806 | 85 | |
| GenBank mRNA | JI895752.1 | - | 7 - 8 | 778 | 85 | |
| JP048768.1 | - | 7 - 8 | 778 | 85 | ||
More Information
Set Membership: |
|

Wbp2
Genomic Map