Assay Details
| Gene Symbol: Oat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507286.2 | XP_003507334.1 | 5 - 6 | 1109 | 78 | |
| XM_007647853.1 | XP_007646043.1 | 5 - 6 | 1113 | 78 | ||
More Information
Set Membership: |
|

Oat
Genomic Map