Assay Details
| Gene Symbol: Dnajc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504638.2 | XP_003504686.1 | 5 - 6 | 555 | 91 | |
| GenBank mRNA | JI876125.1 | - | 5 - 6 | 502 | 91 | |
| JP061831.1 | - | 5 - 6 | 502 | 91 | ||
More Information
Set Membership: |
|

Dnajc7
Genomic Map