Assay Details
| Gene Symbol: Pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007617281.1 | XP_007615471.1 | 5 - 6 | 748 | 56 | |
| GenBank mRNA | JP056228.1 | - | 5 - 6 | 748 | 56 | |
More Information
Set Membership: |
|

Pi4k2a
Genomic Map