Assay Details
Gene Symbol: Grm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498096.2 | XP_003498144.1 | 2 - 3 | 737 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
