Assay Details
| Gene Symbol: Fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498301.2 | XP_003498349.2 | 5 - 6 | 699 | 79 | |
| XM_007625982.1 | XP_007624172.1 | 5 - 6 | 720 | 79 | ||
More Information
Set Membership: |
|

Fam168b
Genomic Map