Assay Details
| Gene Symbol: Cops3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503344.2 | XP_003503392.2 | 2 - 3 | 384 | 81 | |
| GenBank mRNA | JI891568.1 | - | 2 - 3 | 384 | 81 | |
| JP054293.1 | - | 2 - 3 | 384 | 81 | ||
More Information
Set Membership: |
|

Cops3
Genomic Map