Assay Details
| Gene Symbol: Sage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640578.1 | XP_007638768.1 | 6 - 7 | 934 | 98 | |
| GenBank mRNA | JI893420.1 | - | 2 - 3 | 233 | 98 | |
| JP044505.1 | - | 2 - 3 | 233 | 98 | ||
More Information
Set Membership: |
|

Sage1
Genomic Map