Assay Details
| Gene Symbol: Rac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649008.1 | XP_007647198.1 | 4 - 5 | 577 | 81 | |
| GenBank mRNA | JI869845.1 | - | 4 - 5 | 361 | 81 | |
More Information
Set Membership: |
|

Rac1
Genomic Map