Assay Details
| Gene Symbol: Dusp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501260.2 | XP_003501308.1 | 2 - 3 | 728 | 73 | |
| GenBank mRNA | JI895734.1 | - | 2 - 3 | 555 | 73 | |
More Information
Set Membership: |
|

Dusp16
Genomic Map