Assay Details
| Gene Symbol: Syncrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495627.2 | XP_003495675.1 | 9 - 10 | 1309 | 76 | |
| XM_003495628.2 | XP_003495676.1 | 9 - 10 | 1311 | 76 | ||
| XM_007642761.1 | XP_007640951.1 | 9 - 10 | 1309 | 76 | ||
| GenBank mRNA | JI886471.1 | - | 8 - 9 | 1132 | 76 | |
| JP038519.1 | - | 8 - 9 | 1128 | 76 | ||
More Information
Set Membership: |
|

Syncrip
Genomic Map