Assay Details
| Gene Symbol: Ccnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496026.2 | XP_003496074.1 | 4 - 5 | 1018 | 57 | |
| GenBank mRNA | JI876964.1 | - | 4 - 5 | 768 | 57 | |
| JP060574.1 | - | 4 - 5 | 768 | 57 | ||
More Information
Set Membership: |
|

Ccnd2
Genomic Map