Assay Details
| Gene Symbol: Zfand5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513945.2 | XP_003513993.1 | 4 - 5 | 3602 | 102 | |
| GenBank mRNA | JI888860.1 | - | 4 - 5 | 519 | 102 | |
| JP061677.1 | - | 4 - 5 | 519 | 102 | ||
More Information
Set Membership: |
|

Zfand5
Genomic Map