Assay Details
| Gene Symbol: Ctsz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496747.2 | XP_003496795.1 | 3 - 4 | 608 | 58 | |
| GenBank mRNA | AJ303074.1 | - | 3 - 4 | 587 | 58 | |
| JI881997.1 | - | 3 - 4 | 582 | 58 | ||
| JP060604.1 | - | 3 - 4 | 582 | 58 | ||
More Information
Set Membership: |
|

Ctsz
Genomic Map