Assay Details
| Gene Symbol: Klf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642710.1 | XP_007640900.1 | 1 - 2 | 536 | 74 | |
| XM_007642711.1 | XP_007640901.1 | 2 - 3 | 325 | 74 | ||
| XM_007642712.1 | XP_007640902.1 | 2 - 3 | 337 | 74 | ||
| XM_007642713.1 | XP_007640903.1 | 2 - 3 | 377 | 74 | ||
| XM_007642714.1 | XP_007640904.1 | 1 - 2 | 536 | 74 | ||
| XM_007642715.1 | XP_007640905.1 | 1 - 2 | 536 | 74 | ||
More Information
Set Membership: |
|

Klf7
Genomic Map