Assay Details
| Gene Symbol: Smad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502947.2 | XP_003502995.1 | 4 - 5 | 1176 | 72 | |
| GenBank mRNA | JI868294.1 | - | 1 - 2 | 115 | 72 | |
| JP061315.1 | - | 1 - 2 | 115 | 72 | ||
More Information
Set Membership: |
|

Smad9
Genomic Map