Assay Details
| Gene Symbol: Arsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514046.2 | XP_003514094.1 | 6 - 7 | 1525 | 66 | |
| GenBank mRNA | JI879986.1 | - | 6 - 7 | 1477 | 66 | |
| JP037420.1 | - | 6 - 7 | 1477 | 66 | ||
More Information
Set Membership: |
|

Arsa
Genomic Map