Assay Details
| Gene Symbol: Pcf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647544.1 | XP_007645734.1 | 13 - 14 | 4647 | 106 | |
| GenBank mRNA | JI892738.1 | - | 6 - 7 | 749 | 106 | |
More Information
Set Membership: |
|

Pcf11
Genomic Map