Assay Details
| Gene Symbol: Klf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494905.2 | XP_003494953.1 | 1 - 2 | 334 | 62 | |
| GenBank mRNA | JI893687.1 | - | 1 - 2 | 327 | 62 | |
| JP041355.1 | - | 1 - 2 | 327 | 62 | ||
More Information
Set Membership: |
|

Klf6
Genomic Map