Assay Details
| Gene Symbol: Trappc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497176.2 | XP_003497224.1 | 3 - 4 | 272 | 67 | |
| GenBank mRNA | JP063858.1 | - | 3 - 4 | 272 | 67 | |
More Information
Set Membership: |
|

Trappc1
Genomic Map