Assay Details
| Gene Symbol: Ppm1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502845.2 | XP_003502893.1 | 7 - 8 | 1094 | 57 | |
| GenBank mRNA | JI884520.1 | - | 2 - 3 | 153 | 57 | |
| JP049700.1 | - | 2 - 3 | 153 | 57 | ||
More Information
Set Membership: |
|

Ppm1f
Genomic Map