Assay Details
| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642482.1 | XP_007640672.1 | 6 - 7 | 603 | 116 | |
| XM_007642483.1 | XP_007640673.1 | 6 - 7 | 652 | 116 | ||
| GenBank mRNA | JI881198.1 | - | 4 - 5 | 470 | 116 | |
| JP044755.1 | - | 4 - 5 | 470 | 116 | ||
More Information
Set Membership: |
|

Ctdp1
Genomic Map