Assay Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509553.2 | XP_003509601.1 | 11 - 12 | 1021 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mapk12| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509553.2 | XP_003509601.1 | 11 - 12 | 1021 | 94 | |
Set Membership: |
|